missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAF13 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24672-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Initiation of transcription by RNA polymerase II requires the activities of more than 70 polypeptides. The protein that coordinates these activities is transcription factor IID (TFIID), which binds to the core promoter to position the polymerase properly, serves as the scaffold for assembly of the remainder of the transcription complex, and acts as a channel for regulatory signals. TFIID is composed of the TATA-binding protein (TBP) and a group of evolutionarily conserved proteins known as TBP-associated factors or TAFs. TAFs may participate in basal transcription, serve as coactivators, function in promoter recognition or modify general transcription factors (GTFs) to facilitate complex assembly and transcription initiation. This gene encodes a small subunit associated with a subset of TFIID complexes. This subunit interacts with TBP and with two other small subunits of TFIID, TAF10 and TAF11. There is a pseudogene located on chromosome 6.Specifications
TAF13 | |
Polyclonal | |
Unconjugated | |
TAF13 | |
TAF(II)18, TAF13, TAF2K, TAFII 18, TAFII18 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
310784, 6884, 99730 | |
-20°C | |
Liquid |
Western Blot | |
0.6 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P61216, Q15543 | |
Taf13 | |
TAF13 Fusion Protein Ag18193 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |