missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAF1B Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12818-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
Component of RNA polymerase I core factor complex that acts as a GTF2B/TFIIB-like factor and plays a key role in multiple steps during trancription initiation such as preinitiation complex (PIC) assembly and postpolymerase recruitment events in polymerase I (Pol I) transcription. Binds rDNA promoters and plays a role in Pol I recruitment as a component of the SL1/TIF-IB complex and, possibly, directly through its interaction with RRN3.Specifications
TAF1B | |
Polyclonal | |
Unconjugated | |
TAF1B | |
RAF1B, RAFI63, SL1, TAF1B, TAFI63, TBP associated factor 1B | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
9014 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q53T94 | |
TAF1B | |
TAF1B Fusion Protein Ag3267 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |