missing translation for 'onlineSavingsMsg'
Läs mer

Invitrogen™ TAF5 Polyclonal Antibody
GREENER_CHOICE

Produktkod. 16531212
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 μL
Förpackningsstorlek:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16531212 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 16531212 Leverantör Invitrogen™ Leverantörsnummer PA598386

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

Denna artikel kan inte returneras. Se returpolicy

Rabbit Polyclonal Antibody

TAF5 Polyclonal Antibody for Western Blot, IHC (P), ELISA

TFIID is a general transcription factor which initiates preinitiation complex assembly through direct interaction with the TATA promoter element. It is a multisubunit complex consisting of a small TATA-binding polypeptide andother TATA-binding protein (TBP)-associated factors (TAFs). Although native TFIID can mediate both activator-independent (basal) and activator-dependent transcription in reconstituted systems, TBP can mediate only basal transcription. TAF II p100 (TBP-associated factor II100), also known as TAF5 or TAFII100, is the third largest subunit of human TFIID. It contains six WD40 repeats at the C-terminus and has an N-terminus capable of forming a flexible dimer. TAF II p100 plays an important role in forming the scaffold that is crucial for the assembly of the TFIID complex. TAF II p100 may also be involved in the stabilization of TAF interactions.
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen TAF5
Användningsområden ELISA, Immunohistochemistry (Paraffin), Western Blot
Klassificering Polyclonal
Koncentration 0.35 mg/mL
Konjugera Unconjugated
Formulering PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gen TAF5
Genaccessionsnr. Q15542
Gene Alias 6330528C20Rik; AV117817; TAF(II)100; TAF2D; TAF5; TAF5 RNA polymerase II, TATA box binding protein (TBP)-associated factor; TAF5 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 100kDa; TAFII100; TAFII-100; TATA box binding protein (TBP)-associated factor 2D; TATA box binding protein (TBP)-associated factor, RNA polymerase II, D, 100kD; TATA box binding protein associated factor 5; TATA-box binding protein associated factor 5; Tbp-associated factor 5; transcription initiation factor TFIID 100 kD subunit; transcription initiation factor TFIID 100 kDa subunit; Transcription initiation factor TFIID subunit 5
Gensymboler TAF5
Värd art Rabbit
Immunogen Recombinant Human Transcription initiation factor TFIID subunit 5 protein (656-800AA).
Reningsmetod Antigen affinity chromatography
Kvantitet 100 μL
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 6877
Målarter Human
Innehåll och lagring -20°C or -80°C if preferred
Produkttyp Antibody
Form Liquid
Isotyp IgG
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.