missing translation for 'onlineSavingsMsg'
Läs mer

Invitrogen™ TAFII p135/p105 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA536243

Produktkod. 15577581

  • 4595.00 SEK / 100µL

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

Denna artikel kan inte returneras. Se returpolicy

Beskrivning

Beskrivning

This antibody detects endogenous protein at a molecular weight of 135 kDa and 105 kDa. Purity is >95% by SDS-PAGE.

TAF II p105, also called TAF4B, is a celltype specific transcriptional co-activator that is a component of the TFIID complex. Expressed primarily in B cells and ovarian granulosa cells, TAF II p105 can interact with OCBA/POU2AF1 to activate B cell-specific octamerdependent transcription. Additionally, TAF II p105 plays an important role in co-activating the transcription factor NFkB and is essential for activation of anti-apoptotic genes such as TNFAIP3. TAF II p135, also known as TAF4, TAF2C, TAF2C1, TAF4A or TAFII130, is a 1,085 amino acid subunit of TFIID that accelerates transcriptional activation triggered by thyroid hormone (TR) or retinoic acid (RA). Localized to the nucleus, TAF II p135 contains one TAFH domain and is thought to bind to proteins that contain glutamine-rich domains, such as the transcription factor CREB.
TRUSTED_SUSTAINABILITY
Specifikationer

Specifikationer

TAFII p135/p105
Polyclonal
Unconjugated
TAF4
105kDa; 135kDa; 2610524B04Rik; 4932409F03Rik; AI450312; AW987595; RGD1562997; RNA polymerase II TBP-associated factor subunit C; similar to Transcription initiation factor TFIID 105 kDa subunit (TAFII-105); SPGF13; TAF(II)105; TAF(II)130; TAF(II)135; TAF2C; TAF2C1; Taf2c2; TAF4; TAF4 RNA polymerase II, TATA box binding protein (TBP)-associated factor; TAF4 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 135kDa; Taf4a; TAF4A RNA polymerase II, TATA box binding protein (TBP)-associated factor; TAF4A RNA polymerase II, TATA box binding protein (TBP)-associated factor, 135 kD; Taf4b; TAF4B RNA polymerase II, TATA box binding protein (TBP)-associated factor; TAF4B RNA polymerase II, TATA box binding protein (TBP)-associated factor, 105 kDa; TAF4b RNA polymerase II, TATA box binding protein (TBP)-associated factor, 105kDa; TAFII 130; TAFII 135; TAFII105; TAFII-105; TAFII130; TAFII-130; TAFII135; TAFII-135; TATA box binding protein (TBP)-associated factor 4B; TATA box binding protein (TBP)-associated factor, RNA polymerase II, C1, 130kd; TATA box binding protein (TBP)-associated factor, RNA polymerase II, C2, 105kD; TATA box binding protein associated factor 4; TATA box binding protein associated factor 4b; TATA-box binding protein associated factor 4; TATA-box binding protein associated factor 4b; TBP-associated factor 4; Transcription initiation factor TFIID 105 kDa subunit; transcription initiation factor TFIID 130 kDa subunit; transcription initiation factor TFIID 135 kD subunit; transcription initiation factor TFIID 135 kDa subunit; Transcription initiation factor TFIID subunit 4; transcription initiation factor TFIID subunit 4B
Rabbit
Antigen affinity chromatography
RUO
228980, 291773, 682097, 6874, 6875, 72504
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
G5E8Z2, O00268, Q92750
TAF4, TAF4B
Synthetic peptide corresponding to amino acids 951-1000 of human TAF II p135.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Produktförslag

Produktförslag

Filmer
SDS
Dokument

Dokument

Certifikat
Kampanjer

Kampanjer

Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.