Learn More
TAGLN3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12246-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This antibody was generated against full length transgenlin 3 protein. It may cross-react with other two transgenlins based on the sequence similarity.
The transgelin family is a group of proteins that belong to 22 kDa actin-related corpnin superfamily. Of all three isoforms, transgelin 1 is the best characterized. Transgelin 1, also known as SM22alpha, is a specific marker for differentiated smooth muscle cells. Transgelin 2, also known as SM22 beta, is expressed by both smooth muscle and non-smooth muscle cells in a temporally and spatially regulated pattern. Trangenlin 3, also known as NP25, is only found in highly differentiated neuronal cells.Specifications
TAGLN3 | |
Polyclonal | |
Unconjugated | |
TAGLN3 | |
Neuronal protein 22, Neuronal protein NP25, TAGLN3, transgelin 3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
29114, 56370, 63837 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
0.28 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P37805, Q9R1Q8, Q9UI15 | |
TAGLN3 | |
TAGLN3 Fusion Protein Ag2891 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |