Learn More
TAIP12 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA564982
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: GVCILEEPLAV PEELCPGLTA PILIQAQLPP GSSVLCFTEN SDHPTASTVN SPSYLNSGPL VYYQVEQRPV LGVKG Highest antigen sequence identity to the following orthologs - mouse 88%, rat 88%.
The protein encoded by this gene belongs to the CSRNP family of nuclear proteins that share conserved regions, including cysteine- and serine- rich regions, a basic domain, a transcriptional activation domain, and bind the sequence 'AGAGTG', thus have the hallmark of transcription factors. Studies in mice suggest that these genes may have redundant functions. Alternatively spliced transcript variants have been found for this gene.Specifications
TAIP12 | |
Polyclonal | |
Unconjugated | |
Csrnp2 | |
BC035295; C12orf2; C12orf22; Csnrp2; CSRN2; Csrnp2; CSRNP-2; cysteine and serine rich nuclear protein 2; cysteine/serine-rich nuclear protein 2; cysteine-serine-rich nuclear protein 2; Fam130a1; family with sequence similarity 130, member A1; FLJ25576; PPP1R72; Protein FAM130A1; protein phosphatase 1, regulatory subunit 72; RGD1308916; TAIP12; TAIP-12; TGF-beta induced apoptosis protein 12; TGF-beta-induced apoptosis protein 12 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
81566 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H175 | |
Csrnp2 | |
Recombinant Human TAIP12 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only