Läs mer
Tocris Bioscience™ TAK 21d
Potent FAAH inhibitor
Brand: Tocris Bioscience™ 5209/50
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
TAK 21d is a potent FAAH inhibitor; (IC50 values are 0.28 and 0.72 nM, at rat and human FAAH respectively). Displays analgesic effects in vivo models of neuropathic and inflammatory pain. Brain penetrant.Specifikationer
| 4-[4-(3,4-Difluorophenyl)-2-pyrimidinyl]-N-3-pyridazinyl-1-piperazinecarboxamide | |
| 1143578-94-2 | |
| 50 mg |