Läs mer
Tocris Bioscience™ TAK 960 hydrochloride
Beskrivning
TAK 960 hydrochloride is a potent PLK1 inhibitor (IC50 values are 0.8, 16.9 and 50.2 nM for PLK1, PLK2 and PLK3 respectively). Exhibits >20-fold selectivity for PLK1 over FAK, MLCK and FES, and has minimal activity against a panel of 282 other kinases. Inhibits proliferation of a range of cancer cell lines in vitro and inhibits tumor growth of multiple human cancer cell xenografts in vivo. Orally bioavailable.
Specifikationer
Specifikationer
| CAS | 2108449-45-0 |
| Kvantitet | 10 mg |
| Produkttyp | TAK 960 hydrochloride |
| Renhet | >98% |
| Molekylvikt (g/mol) | 598.06 |
For Research Use Only