Läs mer
Talabostat mesylate, MedChemExpress
Produktkod. 16478267
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
Förpackningsstorlek:
10mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16478267 5 mg 5mg
16405578 10 mg 10mg
16481737 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 16478267 Leverantör Medchem Express Leverantörsnummer HY13233A
MedChemExpress Talabostat mesylate (Val-boroPro mesylate; PT100 mesylate) is an orally active and nonselective dipeptidyl peptidase IV (DPP-IV) inhibitor (IC50 Ki = 0.18 nM) and the first clinical inhibitor of fibroblast activation protein (FAP) (IC50 = 560 nM), inhibits DPP8/9 (IC50 = 4/11 nM; Ki = 1.5/0.76 nM), quiescent cell proline dipeptidase (QPP) (IC50 = 310 nM), DPP2, and some other DASH family enzymes.
Antineoplastic and hematopoiesis- stimulating activities.
Specifikationer
Kemiskt namn eller material Talabostat mesylate
CAS 150080-09-4
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C10H23BN2O6S
Kvantitet 5 mg
Synonym Val-boroPro mesylatePT100 mesylate
Löslighetsinformation H2O : 250 mg/mL (805.98 mM; Need ultrasonic) ∣DMSO : ≥ 40 mg/mL (128.96 mM)
LEDER CC(C)[C@H](N)C(N1[C@H](B(O)O)CCC1)=O.CS(=O)(O)=O
Molekylvikt (g/mol) 310.18
Formel vikt 310.18
Procent renhet 95.0%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Cancer-Kinase/protease