Läs mer
Talaporfin sodium, MedChemExpress
Produktkod. 16477058
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16477058 10 mg
16439607 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 16477058 Leverantör Medchem Express Leverantörsnummer HY16477
MedChemExpress Talaporfin (ME2906; NPe6) is a photosensitizer used in photodynamic therapy (PDT).
Talaporfin (ME2906
Specifikationer
Kemiskt namn eller material Talaporfin sodium
Fysisk form Powder
CAS 220201-34-3
Färg Black
Kvantitet 10 mg
Anteckningar om renhetsgrad Research
Molekylformel C38H37N5Na4O9
Synonym ME2906 Mono-L-aspartyl chlorin e6 NPe6
Löslighetsinformation H2O : 25 mg/mL (31.26 mM; Need ultrasonic) ∣DMSO : 2 mg/mL (2.50 mM; Need ultrasonic)
LEDER O=C(O[Na])C[C@@H](C(O[Na])=O)NC(C/C1=C2[C@@H](CCC(O[Na])=O)[C@H](C)C(/C=C3C(C)=C(C=C)/C(N/3)=C/C(C(C)=C/4CC)=NC4=C/C5=C(C)C(C(O[Na])=O)=C1N5)=N\2)=O
Molekylvikt (g/mol) 799.69
Formel vikt 799.69
Procent renhet 95.0%