Läs mer
Taminadenant, MedChemExpress
Produktkod. 16447007
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16447007 5 mg
16430617 50 mg
16424288 10 mg
16405216 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16447007 Leverantör Medchem Express Leverantörsnummer HY109139
MedChemExpress Taminadenant is an antagonist of adenosine receptor.
Taminadenant is an antagonist of adenosine receptor.
Specifikationer
Kemiskt namn eller material Taminadenant
Fysisk form Solid
CAS 1337962-47-6
Färg White
Kvantitet 5 mg
Anteckningar om renhetsgrad Research
Molekylformel C10H8BrN7
Löslighetsinformation DMSO : 125 mg/mL (408.34 mM; Need ultrasonic)
LEDER BrC(C(N1C=CC=N1)=NC(N2C=CC=N2)=N3)=C3N
Molekylvikt (g/mol) 306.12
Formel vikt 306.12
Procent renhet 99.13%