Läs mer
Tamoxifen Citrate EP Impurity F (as Hydrochloride), Mikromol™
Produktkod. 30051393
Change view
Kvantitet:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30051393 25 mg
30049455 100 mg
30461137 4 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30051393 Leverantör Mikromol Leverantörsnummer MM0053.120025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Tamoxifen Citrate EP Impurity F (as Hydrochloride)
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-[4-[(Z)-1,2-Diphenylbut-1-enyl]phenoxy]-N-methylethanamine Hydrochloride
CAS 15917-65-4
Analyt- eller komponentnamn 2-[4-[(Z)-1,2-Diphenylbut-1-enyl]phenoxy]-N-methylethanamine Hydrochloride
Frakt skick Room Temperature
Molekylformel C25 H27 N O . Cl H
Certifikat och efterlevnad ISO 17025
Synonym 2-[4-[(Z)-1,2-Diphenylbut-1-enyl]phenoxy]-N-methylethanamine Hydrochloride, Tamoxifen Citrate Imp. F (EP) as Hydrochloride, Tamoxifen Citrate Impurity F as Hydrochloride, Tamoxifen Impurity F as Hydrochloride
InChI formel InChI=1S/C25H27NO.ClH/c1-3-24(20-10-6-4-7-11-20)25(21-12-8-5-9-13-21)22-14-16-23(17-15-22)27-19-18-26-2;/h4-17,26H,3,18-19H2,1-2H3;1H/b25-24-;
LEDER Cl.CC\C(=C(/c1ccccc1)\c2ccc(OCCNC)cc2)\c3ccccc3
IUPAC-namn 2-[4-[(Z)-1,2-diphenylbut-1-enyl]phenoxy]-N-methylethanamine;hydrochloride
Molekylvikt (g/mol) 393.95
Kvantitet 25 mg
Formel vikt 393.1859 g/mol