Läs mer
Tocris Bioscience™ Tandospirone hydrochloride
GREENER_CHOICE
Produktkod. 18015242
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18015242 10 mg
18014052 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18015242 Leverantör Tocris Bioscience™ Leverantörsnummer 2854/10
Selective 5-HT1A partial agonist
5-HT1A receptor partial agonist (Ki = 27 nM) that displays selectivity over 5-HT2, 5-HT1C, α1, α2, D1 and D2 receptors (Ki values ranging from 1300 - 41000 nM). Inactive at 5-HT uptake sites, 5-HT1B, β-adrenergic, muscarinic and benzodiazepine receptors. Displays anxiolytic activity.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 99095-10-0
Molekylformel C21H30ClN5O2
Specifikationer
Kemiskt namn eller material Tandospirone hydrochloride
Kvantitet 10 mg
Formel vikt 419.95
Renhet >98%
Molekylvikt (g/mol) 419.95