Learn More
Corning™ Tank Liners
Product Code. 31932521 Shop All Corning Products
Change view
Click to view available options
Capacity (Metric):
560 L
Unit Size:
Pack of 20
Product Code. Capacity (Metric) Unit Size
31932521 560 L Pack of 20
1 options
Product Code. 31932521 Supplier Corning™ Supplier No. 9130060
Beräknad leverans: 30-12-2026
Designed to fit standardized cylindrical tanks manufactured by Nalge™/Nunc™ and other brands
  • Made from low-density polyethylene (LDPE)
  • Eliminate costs associated with cleaning and validation of tanks
  • Decrease labor costs and increase turnaround time
  • Eliminate risks associated with cross contamination
  • Help extend life of reuseable tanks, resulting in reduced capital expenditures
  • Gamma irradiated, individually packaged
  • Choice of gusseted and non-gusseted available
  • Available in many sizes
TRUSTED_SUSTAINABILITY
Specifications
Material LDPE
Product Type Non-Gusseted Tank Liner
Capacity (Metric) 560 L