missing translation for 'onlineSavingsMsg'
Learn More

Tannic acid

900.00 SEK - 1625.00 SEK

Chemical Identifiers

CAS 637-01-4
Molecular Formula C10H18Cl2N2
Molecular Weight (g/mol) 237.168
InChI Key FBHKTSXMTASXFJ-UHFFFAOYSA-N
Synonym n,n,n',n'-tetramethyl-p-phenylenediamine dihydrochloride, wurster's reagent dihydrochloride, n,n,n',n'-tetramethyl-1,4-phenylenediamine dihydrochloride, unii-66w8hka51x, n1,n1,n4,n4-tetramethylbenzene-1,4-diamine dihydrochloride, wurster's blue dihydrochloride, 1,4-benzenediamine, n,n,n',n'-tetramethyl-, dihydrochloride, 1,4-benzenediamine, n1,n1,n4,n4-tetramethyl-, hydrochloride 1:2, 1-n,1-n,4-n,4-n-tetramethylbenzene-1,4-diamine dihydrochloride, wursters reagent
PubChem CID 71561
IUPAC Name 1-N,1-N,4-N,4-N-tetramethylbenzene-1,4-diamine;dihydrochloride
SMILES CN(C)C1=CC=C(C=C1)N(C)C.Cl.Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11491781
View Documents
MP Biomedicals
0219027590
500g 900.00 SEK
500g View alternative products
Save Up to 
»
Estimated Shipment
09-09-2020
Add to basket
 
11401791
View Documents
MP Biomedicals
0219027591
1kg 1625.00 SEK
1kg View alternative products
Save Up to 
»
Estimated Shipment
09-09-2020
Add to basket
 
Specifications

Chemical Identifiers

637-01-4
237.168
n,n,n',n'-tetramethyl-p-phenylenediamine dihydrochloride, wurster's reagent dihydrochloride, n,n,n',n'-tetramethyl-1,4-phenylenediamine dihydrochloride, unii-66w8hka51x, n1,n1,n4,n4-tetramethylbenzene-1,4-diamine dihydrochloride, wurster's blue dihydrochloride, 1,4-benzenediamine, n,n,n',n'-tetramethyl-, dihydrochloride, 1,4-benzenediamine, n1,n1,n4,n4-tetramethyl-, hydrochloride 1:2, 1-n,1-n,4-n,4-n-tetramethylbenzene-1,4-diamine dihydrochloride, wursters reagent
1-N,1-N,4-N,4-N-tetramethylbenzene-1,4-diamine;dihydrochloride
C10H18Cl2N2
FBHKTSXMTASXFJ-UHFFFAOYSA-N
71561
CN(C)C1=CC=C(C=C1)N(C)C.Cl.Cl

Specifications

Tannic Acid
C10H18Cl2N2
FBHKTSXMTASXFJ-UHFFFAOYSA-N
1-N,1-N,4-N,4-N-tetramethylbenzene-1,4-diamine;dihydrochloride
71561
Yellow
637-01-4
n,n,n',n'-tetramethyl-p-phenylenediamine dihydrochloride, wurster's reagent dihydrochloride, n,n,n',n'-tetramethyl-1,4-phenylenediamine dihydrochloride, unii-66w8hka51x, n1,n1,n4,n4-tetramethylbenzene-1,4-diamine dihydrochloride, wurster's blue dihydrochloride, 1,4-benzenediamine, n,n,n',n'-tetramethyl-, dihydrochloride, 1,4-benzenediamine, n1,n1,n4,n4-tetramethyl-, hydrochloride 1:2, 1-n,1-n,4-n,4-n-tetramethylbenzene-1,4-diamine dihydrochloride, wursters reagent
CN(C)C1=CC=C(C=C1)N(C)C.Cl.Cl
237.168
Powder
500g