Läs mer
Tanomastat, TRC
Produktkod. 30415729
Change view
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30415729 50 mg
30413425 5 mg
30417249 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30415729 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT00690050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 179545-77-8
Molekylformel C23 H19 Cl O3 S
Molekylvikt (g/mol) 410.91
IUPAC-namn (2S)-4-[4-(4-chlorophenyl)phenyl]-4-oxo-2-(phenylsulfanylmethyl)butanoic acid
LEDER OC(=O)[C@@H](CSc1ccccc1)CC(=O)c2ccc(cc2)c3ccc(Cl)cc3
Specifikationer
Kemiskt namn eller material Tanomastat
Kvantitet 50 mg
InChI formel InChI=1S/C23H19ClO3S/c24-20-12-10-17(11-13-20)16-6-8-18(9-7-16)22(25)14-19(23(26)27)15-28-21-4-2-1-3-5-21/h1-13,19H,14-15H2,(H,26,27)/t19-/m1/s1
Formel vikt 410.0743