Learn More
TAP26 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA558439
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: NLKHLYLAEE ERHRKQARKV DHPLSEQVHQ PLLEEQCSID EPLFEDQCSF DQPQPEEQCI KTVNSFTIPK K Highest antigen sequence identity to the following orthologs: Mouse - 53%, Rat - 61%.
Component of the transcription complexes of the pulmonary surfactant-associated protein-B (SFTPB) and -C (SFTPC). Enhances homeobox protein Nkx-2.1-activated SFTPB and SFTPC promoter activities.Specifications
TAP26 | |
Polyclonal | |
Unconjugated | |
CCDC59 | |
2300004H16Rik; BR22; Ccdc59; coiled-coil domain containing 59; coiled-coil domain-containing protein 59; D10Ertd718e; HSPC128; RGD1311912; TAP26; Thyroid transcription factor 1-associated protein 26; thyroid transcription factor 1-associated protein 26-like protein; TTF-1-associated protein 26; TTF-1-associated protein BR2; Unknown (protein for IMAGE:8047375) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
29080 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9P031 | |
CCDC59 | |
Recombinant protein corresponding to Human TAP26 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only