Läs mer
Micronova™ Tape Controlled Environment
Produktkod. 13138515 Handla allt Micronova Produkter
Change view
Färg:
Orange
Förpackningsstorlek:
Varje
Produktkod. Färg unitSize
13138515 Orange Varje
1 options
Produktkod. 13138515 Leverantör Micronova™ Leverantörsnummer PCX3OR
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
PCX™ Cleanroom tape can be used for colour-coding, sealing, labelling and masking, and various tasks within the cleanroom requiring pressure-sensitive qualities
polyethylene, orange 76mm x 33m Premium
Specifikationer
Färg Orange
Formatera Roll
Material Polyethylene
Sterilitet Non-sterile
För användning med (applikation) Cleanroom use - color-coding, sealing, labelling and masking, or for pressure-sensitive applications
Längd (metrisk) 33 m
Bredd (metrisk) 7.6 cm