missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAPT1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA565524
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: TLNVAFNSHNK SLLTIMMSNN FVEIKGSVFK KFEKNNLFQM SNSDIKERFT NYVLLLIVCL RNMEQFSWNP DHL Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.
TAPT1 Polyclonal antibody specifically detects TAPT1 in Human samples. It is validated for Immunocytochemistry.Specifications
TAPT1 | |
Polyclonal | |
Unconjugated | |
TAPT1 | |
4932414K18Rik; CMVFR; Cytomegalovirus partial fusion receptor; OCLSBG; TAPT1; transmembrane anterior posterior transformation 1; transmembrane anterior posterior transformation protein 1; Transmembrane anterior posterior transformation protein 1 homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
202018 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6NXT6 | |
TAPT1 | |
Recombinant Human TAPT1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only