Learn More
MP Biomedicals™ Taq Pol Bulk
Product Code. 11497010 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 x 250U
1000U
250U
3 x 250U
5 x 1000U
Unit Size:
1000 units
250 units
750 units
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11497010 3 x 250U 750 units
11467010 250U 250 units
11477010 1000U 1000 units
11417020 10 x 250U 250 units
11487010 5 x 1000U 1000 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11497010 Supplier MP Biomedicals™ Supplier No. 11EPTQA325
  • ONLINE EXCLUSIVE PRICE
    5975.00 SEK
    5690.27 SEK / Paket med 750
    Save 284.73 SEK
    4% Av
 Restricted Item
Beräknad leverans: 26-11-2026
For life science applications
  • Ready for CE IVD labeling of clinical applications
Specifications
Concentration 5 U/μL
Description Taq Pol Bulk
Quantity 3 x 250U
Includes Taq DNA Polymerase, 10X Standard Buffer with Mg, 10X Standard Buffer without Mg, 25mM MgCl2
For Resaerch Use Only