Learn More
Applied Biosystems™ TaqMan™ Array Human TNF Superfamily Pathway
Brand:  Applied Biosystems™ 4414189
Additional Details : Weight : 0.00450kg
Product Code. 10239334
  • 2250.00 SEK / 1 set
Estimated Shipment: 03-06-2024
to see stock.
For Research Use Only. All usage must comply with product instructions.
Description
Description
Manual transfer of reagents into plates during routine experimental setups can be time-consuming and tedious. Daily preparation of such plates can negatively impact overall productivity. To ease the stress and minimize the error rate, TaqMan™ Array Plates are designed to reduce redundant pipetting steps and minimize loading errors, making them an ideal solution for fast and easy experimental setup. TaqMan™ Array Plates contain high-quality TaqMan™ Gene Expression Assays (TaqMan™ probe and PCR primer sets) in a convenient, dried-down, 96- or 384-well plate format. Extensive tests have found no difference in performance between TaqMan™ Assays supplied in solution in single tubes and assays supplied dried in a TaqMan™ Array Plate
  • Extensive collections - choose from over 130 predesigned TaqMan™ Gene Signature Arrays and Plates, or customize your own plate, selecting from any of our 1.3 million inventoried assays across 24 species
  • Affordable - minimum order of only one gene signature plate or six custom plates
  • Convenient - dried TaqMan probe and primer sets supplied in 96-well plates; just add master mix and sample, and then run
  • Quick results - Fast format typically provides results in under an hour
Specifications
Specifications
Each TaqMan™ Array Plate well contains a single TaqMan™ Assay for one 20 μL reaction. The plates can be stored at room temperature or at 4°C.
96-well plate
MGB (Minor Groove Binder) (3'), Non-Fluorescent Quencher (3'), FAM (5')
TaqMan™
Cell Cycle & Proliferation, Apoptosis, Biomarker-Related Pathway(s), Development, Immunology, Signal Transduction
1 x 20 μL Reactions
Dried Down
Standard
FAM
Standard