Learn More
Applied Biosystems™ TaqMan™ Array Rat Phosphodiesterase Panel
The Applied Biosystems™ TaqMan™ Rat Phosphodiesterase Array is an efficient, easy-to-use micro fluidic card for quantitative gene expression analysis of inflammation genes important in drug discovery.
Brand: Applied Biosystems™ 4378706
11163.00 SEK valid until 2024-09-27
Use promo code "21650" to get your promotional price.
Description
This product is designed with 48 TaqMan™ Gene Expression Assays arrayed in 384 wells and is compatible with an upgraded Applied Biosystems 7900HT Fast Real-Time PCR System and sold in packs of 4.
The array is pre-formatted and inventoried, cost-effective, convenient and easy to use without needing expensive robotics. Results are reproducible and consistent across samples, studies, and labs —the same data quality from card-to-card and lot-to-lot — even with different operators.
Specifications
Each TaqMan™ Array well contains a single TaqMan™ Assay for one 2 μL reaction. The cards should be stored (at 4°C.) | |
384-well microfluidic card | |
MGB (Minor Groove Binder) (3'), Non-Fluorescent Quencher (3'), FAM (5') | |
TaqMan™ | |
Cell Cycle & Proliferation, Oncology, Toxicology & Drug Metabolism, Neurobiology, Inflammation | |
Includes five distinct phosphodiesterase families | |
1 x 2 μL Reactions |
Dried Down | |
Standard | |
FAM | |
4 cards | |
Rat | |
Gene Expression |
For Research Use Only. Not for use in diagnostic procedures.