missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ OpenArray™ Real-Time PCR Master Mix

TaqMan™ OpenArray™ Real-Time PCR Master Mix is optimized for use with TaqMan™ OpenArray™ gene expression panels, and yields reproducible, specific, and sensitive results.

Brand:  Applied Biosystems™ 4462164

Additional Details : Weight : 0.03000kg

 View more versions of this product

Product Code. 15206505

  • 4140.00 SEK / 5mL


In stock, ready to ship: 3
Add to basket

Description

Description

  • Hot start chemistry to provide room temperature stability
  • Optimized for use with TaqMan™ OpenArray™ gene expression panels yielding reproducible, specific, and sensitive results
  • For use with OpenArray™ MicroRNA panels, OpenArray™ gene expression pathway panels, and other gene expression applications on the OpenArray™ format
  • Ready for use with no additional additives required

Order Info

Shipping Conditions: Dry Ice

Specifications

Specifications

Store at -5 to -30°C until first use, then at 2–8°C.
OpenArray®
Tube(s)
34 Panels
TaqMan®
5mL
Total RNA
Target-Specific
Primer-Probe Detection
Liquid
Built-In Hot Start
2-Step qRT-PCR
OpenArray®, TaqMan™
Standard
Dry Ice
Fluorescence Intensity
Videos
Documents

For Research Use Only. Not for use in diagnostic procedures.