Läs mer
Tocris Bioscience™ Targapremir 210
Kvantitet:
5 mg
Beskrivning
Targapremir 210 inhibits formation of mature miR-210 (IC50 = 200 nM) by binding to Dicer binding site of the miR-210 hairpin precursor (Kd = 200 nM) and inhibits Dicer processing of the miRNA. Decreases levels of miR-210 and HIF-1α mRNA, and induces apoptosis in MDA-MB-231 cells cultured under hypoxic conditions. Reduces tumor expression of HIF-1α mRNA and miR-210, and tumor growth in a triple negative breast cancer mouse model.
Specifikationer
Specifikationer
| Kemiskt namn eller material | N-(3-Azidopropyl)-4-[3-[6-(4-methyl-1-piperazinyl)[2,6'-bi-1H-benzimidazol]-2'-yl]phenoxy]butanamide |
| CAS | 1049722-30-6 |
| Kvantitet | 5 mg |
| Molekylformel | C32H36N10O2 |
| Renhet | 98% |