Läs mer
Tocris Bioscience™ Tariquidar dihydrochloride
Beskrivning
Tariquidar dihydrochloride is a potent P-glycoprotein (P-gp) inhibitor (IC50 = 5.1 nM). Reverses drug resistance in multiple MDR cell lines. Acts as a substrate for breast cancer resistance protein (BCRP) at low concentrations and acts as an inhibitor of BCRP when used at >100 nM concentration. Potentiates the cytotoxic effects of doxorubicin (Cat. No. 2252) and mitoxantrone (Cat. No. 4250) in vitro.
Specifikationer
Specifikationer
| Kemiskt namn eller material | N-[2-[[[4-[2-(3,4-dihydro-6,7-dimethoxy-2(1H)-isoquinolinyl)ethyl]phenyl]amino]carbonyl]-4,5-dimethoxyphenyl]-3-quinolinecarbxamide dihydrochloride |
| CAS | 1992047-62-7 |
| Kvantitet | 10 mg |
| Mål | Multidrug Transporter Inhibitors |
| Molekylformel | C38H38N4O6.2HCl |
| Formel vikt | 719.66 |
| Renhet | >98% |