missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TARS2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15067-1-AP-150UL
Additional Details : Weight : 0.00010kg
Specifications
TARS2 | |
Polyclonal | |
Unconjugated | |
TARS2 | |
TARS2, TARSL1, Threonine tRNA ligase, ThrRS | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
310672, 71807, 80222 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q3UQ84, Q68FW7, Q9BW92 | |
Tars2 | |
TARS2 Fusion Protein Ag7074 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |