missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TARSL2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA566861
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: VIPVGPTCEKY ALQVSSEFFE EGFMADVDLD HSCTL Highest antigen sequence identity to the following orthologs - mouse 86%, rat 58%.
TARSL2 Polyclonal antibody specifically detects TARSL2 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Specifications
TARSL2 | |
Polyclonal | |
Unconjugated | |
TARSL2 | |
A530046H20Rik; probable threonine--tRNA ligase 2, cytoplasmic; Probable threonyl-tRNA synthetase 2, cytoplasmic; RGD1311308; si:zfos-508h5.3; TARS3; TARSL2; threonine--tRNA ligase; Threonine--tRNA ligase 2, cytoplasmic; threonine--tRNA ligase 2, cytoplasmic; probable threonine--tRNA ligase 2, cytoplasmic; threonine--tRNA ligase, cytoplasmic; threonyl-tRNA synthetase; threonyl-tRNA synthetase 3; threonyl-tRNA synthetase like 2; threonyl-tRNA synthetase-like 2; threonyl-tRNA synthetase-like protein 2; ThrRS | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
123283 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
A2RTX5 | |
TARSL2 | |
Recombinant Human TARSL2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only