missing translation for 'onlineSavingsMsg'
Learn More

(+)-Tartaric Acid, Certified AR for Analysis, meets analytical specification of Ph.Eur, Fisher Chemical™

Catalog Number p-7073597
Click to view available options
Quantity:
500 g
1 kg
87-69-4
C4H6O6
150.09
MFCD00064207
FEWJPZIEWOKRBE-UHFFFAOYNA-N
l-tartaric acid, l-+-tartaric acid, l +-tartaric acid, 2r,3r-2,3-dihydroxysuccinic acid, tartaric acid, 2r,3r-2,3-dihydroxybutanedioic acid, r,r-tartaric acid, +-l-tartaric acid, dextrotartaric acid, l-threaric acid
444305
CHEBI:15671
(2R,3R)-2,3-dihydroxybutanedioic acid
OC(C(O)C(O)=O)C(O)=O
87-69-4
C4H6O6
150.09
MFCD00064207
FEWJPZIEWOKRBE-UHFFFAOYNA-N
l-tartaric acid, l-+-tartaric acid, l +-tartaric acid, 2r,3r-2,3-dihydroxysuccinic acid, tartaric acid, 2r,3r-2,3-dihydroxybutanedioic acid, r,r-tartaric acid, +-l-tartaric acid, dextrotartaric acid, l-threaric acid
444305
CHEBI:15671
(2R,3R)-2,3-dihydroxybutanedioic acid
OC(C(O)C(O)=O)C(O)=O

(+)-Tartaric Acid, Certified AR for Analysis, meets analytical specification of Ph.Eur

TRUSTED_SUSTAINABILITY
Melting Point 168°C
Color White
pH 1.6
Packaging Plastic powder jar
Quantity 500 g
Formula Weight 150.09g/mol
Vapor Pressure <0.1mbar at 20°C
Physical Form Solid
compliance-icons
Product Identifier
  • L(+)-Tartaric acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'