Läs mer
Tartaric Acid Methyl Ester, TRC
Produktkod. 30423183
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30423183 500 mg
1 options
Produktkod. 30423183 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT007670500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Tartaric Acid Methyl Ester
CAS 3333-46-8
Molekylformel C5H8O6
Kvantitet 500 mg
Synonym (2R,3R)-2,3-Dihydroxy-butanedioic Acid Monomethyl Ester, 2,3-Dihydroxy-[R-(R*,R*)]-butanedioic Acid Monomethyl Ester, Tartaric Acid Monomethyl Ester, (+)-Methyl Hydrogen Tartrate, (2R,3R)-2,3-Dihydroxybutanedioic Acid Monomethyl Ester
InChI formel InChI=1S/C5H8O6/c1-11-5(10)3(7)2(6)4(8)9/h2-3,6-7H,1H3,(H,8,9)/t2-,3-/m1/s1
LEDER COC([C@H](O)[C@@H](O)C(O)=O)=O
IUPAC-namn (2R,3R)-2,3-dihydroxy-4-methoxy-4-oxobutanoic acid
Molekylvikt (g/mol) 164.11
Formel vikt 164.0321