Läs mer
TAS 102 (2:1), TRC
Produktkod. 30410451
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30410451 50 mg
1 options
Produktkod. 30410451 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT00772050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Tipiracil-trifluridine Mixture (2:1)
CAS 733030-01-8
Molekylformel C9H11ClN4O2 • HCl • 2[C10H11F3N2O5]
Kvantitet 50 mg
Synonym Tipiracil-trifluridine Mixture (2:1), trifluridine (TFT) and Tipiracil Hydrochloride (TTP) in molar ratio of 1: 0.5, Trifluridine-tipiracil Hydrochloride Mixture
InChI formel InChI=1S/2C10H11F3N2O5.C9H11ClN4O2.ClH/c2*11-10(12,13)4-2-15(9(19)14-8(4)18)7-1-5(17)6(3-16)20-7;10-7-5(12-9(16)13-8(7)15)4-14-3-1-2-6(14)11;/h2*2,5-7,16-17H,1,3H2,(H,14,18,19);11H,1-4H2,(H2,12,13,15,16);1H/t2*5-,6+,7+;;/m00../s1
LEDER O=C(N1)NC(CN2CCCC2=N)=C(Cl)C1=O.O[C@H]3C[C@H](N(C=C4C(F)(F)F)C(NC4=O)=O)O[C@@H]3CO.O[C@H]5C[C@H](N(C=C6C(F)(F)F)C(NC6=O)=O)O[C@@H]5CO.Cl
IUPAC-namn 1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-(trifluoromethyl)pyrimidine-2,4(1H,3H)-dione--5-chloro-6-((2-iminopyrrolidin-1-yl)methyl)pyrimidine-2,4(1H,3H)-dione (2/1) hydrochloride
Molekylvikt (g/mol) 871.46
Formel vikt 870.16