Läs mer
Tasimelteon, TRC
Produktkod. 17993725
Change view
Kvantitet:
25 mg
100 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17993725 1 g
17992975 100 mg
17943065 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17993725 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT0077401G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 609799-22-6
Molekylformel C15 H19 N O2
Molekylvikt (g/mol) 245.3169
Synonym Propanamide, N-[[(1R,2R)-2-(2,3-dihydro-4-benzofuranyl)cyclopropyl]methyl]-, N-[[(1R,2R)-2-(2,3-Dihydro-4-benzofuranyl)cyclopropyl]methyl]propanamide, (1R-trans)-N-[[2-(2,3-Dihydro-4-benzofuranyl)cyclopropyl]methyl]propanamide, BMS 214778, Hetlioz, MA 1, N-[[(1R,2R)-2-(2,3-Dihydro-1-benzofuran-4-yl)cyclopropyl]methyl]propanamide, Tasimelteon, VEC 162
IUPAC-namn N-[[(1R,2R)-2-(2,3-dihydro-1-benzofuran-4-yl)cyclopropyl]methyl]propanamide
LEDER CCC(=O)NC[C@@H]1C[C@H]1c2cccc3OCCc23
Specifikationer
Kemiskt namn eller material Tasimelteon
Kvantitet 1 g
InChI formel InChI=1S/C15H19NO2/c1-2-15(17)16-9-10-8-13(10)11-4-3-5-14-12(11)6-7-18-14/h3-5,10,13H,2,6-9H2,1H3,(H,16,17)/t10-,13+/m0/s1
Formel vikt 245.1416