Learn More
TASP1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA556567
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: ENDSGTLDTV GAVVVDHEGN VAAAVSSGGL ALKHPGRVGQ AALYGCGCWA ENTGAHNPYS TAVSTSGCGE HLVRTILARE CSHALQAEDA HQAL Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.
TASP1 Polyclonal antibody specifically detects TASP1 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Specifications
TASP1 | |
Polyclonal | |
Unconjugated | |
TASP1 | |
4930485D02Rik; AW986064; C20orf13; dJ585I14.2; FLJ20212; MGC39159; RGD1308591; si:dkey-38k3.3; Tasp1; taspase 1; taspase, threonine aspartase 1; taspase, threonine aspartase, 1; taspase-1; Threonine aspartase 1; Threonine aspartase 1-like protein; Threonine aspartase subunit alpha; Threonine aspartase subunit beta | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55617 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H6P5 | |
TASP1 | |
Recombinant protein corresponding to Human TASP1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only