Läs mer
Tocris Bioscience™ TAT-cyclo-CLLFVY
Selective HIF-1 dimerization inhibitor
Brand: Tocris Bioscience™ 5582/1
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
TAT-cyclo-CLLFVY is a selective HIF-1 dimerization inhibitor. Blocks protein-protein interaction of recombinant HIF-1α, but not HIF-2α, with HIF-1β (IC50 = 1.3 μM) . Inhibits hypoxia-induced HIF-1 activity, and decreases VEGF and CAIX expression in osteosarcoma and breast cancer cells in vitro. Also reduces tubularization of hypoxic HUVECs.Specifikationer
| 95% | |
| Store at -20°C | |
| Observed M.W.: 2559.1 | |
| C111H188N42O24S2 |
| 1446322-66-2 | |
| HIF Inhibitors | |
| 1 mg |