missing translation for 'onlineSavingsMsg'
Läs mer

Taurinamide Hydrochloride, TRC

Produktkod. 30421166
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
2.5 g
Förpackningsstorlek:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30421166 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30421166 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT0078452.5G

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material Taurinamide Hydrochloride
CAS 89756-60-5
Anteckningar om renhetsgrad HPLC
Molekylformel C2 H8 N2 O2 S . HCl
Kvantitet 2.5 g
Synonym 2-Amino-ethanesulfonamide Hydrochloride, 2-Aminoethanesulfonamide Hydrochloride, 2-Sulfamoylethylamine Hydrochloride, Aminoethanesulfonamide Hydrochloride, NSC 403678 Hydrochloride
InChI formel InChI=1S/C2H8N2O2S.ClH/c3-1-2-7(4,5)6;/h1-3H2,(H2,4,5,6);1H
LEDER NCCS(N)(=O)=O.Cl
IUPAC-namn 2-aminoethanesulfonamide;hydrochloride
Molekylvikt (g/mol) 160.62
Formel vikt 160.0073
Procent renhet >95
Visa mer Visa mindre
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.