Läs mer
Taurinamide Hydrochloride, TRC
Produktkod. 30427533
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30427533 250 mg
1 options
Produktkod. 30427533 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT007845250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Taurinamide Hydrochloride
CAS 89756-60-5
Anteckningar om renhetsgrad HPLC
Molekylformel C2 H8 N2 O2 S . HCl
Kvantitet 250 mg
Synonym 2-Amino-ethanesulfonamide Hydrochloride, 2-Aminoethanesulfonamide Hydrochloride, 2-Sulfamoylethylamine Hydrochloride, Aminoethanesulfonamide Hydrochloride, NSC 403678 Hydrochloride
InChI formel InChI=1S/C2H8N2O2S.ClH/c3-1-2-7(4,5)6;/h1-3H2,(H2,4,5,6);1H
LEDER NCCS(N)(=O)=O.Cl
IUPAC-namn 2-aminoethanesulfonamide;hydrochloride
Molekylvikt (g/mol) 160.62
Formel vikt 160.0073