Läs mer
Tauro-Beta-muricholic Acid Sodium Salt, TRC
Produktkod. 30413049
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30413049 25 mg
1 options
Produktkod. 30413049 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT00913525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Tauro-beta-muricholic Acid Sodium Salt
CAS 145022-92-0
Anteckningar om renhetsgrad HPLC
Molekylformel C26 H44 N O7 S . Na
Kvantitet 25 mg
Synonym Tauro-β-muricholic Acid Sodium Salt, Ethanesulfonic acid','2-[[(3α,5β,6β,7β)-3,6,7-trihydroxy-24-oxocholan-24-yl]amino]-','sodium salt (1:1), Ethanesulfonic acid','2-[[(3α,5β,6β,7β)-3,6,7-trihydroxy-24-oxocholan-24-yl]amino]-','monosodium salt (9CI), Sodium tauro-β-muricholate, Tauro-β-muricholic acid sodium salt, 2-[[(3α,5β,6β,7β)-3,6,7-Trihydroxy-24-oxocholan-24-yl]amino]ethanesulfonic acid sodium salt
InChI formel InChI=1S/C26H45NO7S.Na/c1-15(4-7-21(29)27-12-13-35(32,33)34)17-5-6-18-22-19(9-11-25(17,18)2)26(3)10-8-16(28)14-20(26)23(30)24(22)31;/h15-20,22-24,28,30-31H,4-14H2,1-3H3,(H,27,29)(H,32,33,34);/q;+1/p-1/t15-,16-,17-,18+,19+,20+,22+,23+,24-,25-,26-;/m1./s1
LEDER [Na+].C[C@H](CCC(=O)NCCS(=O)(=O)[O-])[C@H]1CC[C@H]2[C@@H]3[C@@H](O)[C@@H](O)[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
IUPAC-namn sodium;2-[[(4R)-4-[(3R,5R,6S,7R,8S,9S,10R,13R,14S,17R)-3,6,7-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]ethanesulfonate
Molekylvikt (g/mol) 537.68
Formel vikt 537.2736