Läs mer
Taurochenodeoxycholic Acid Sodium Salt, TRC
Produktkod. 30137338
Change view
Kvantitet:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137338 500 mg
30133503 1 g
17923855 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137338 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT008130500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 6009-98-9
Molekylformel C26 H44 N O6 S . Na
Molekylvikt (g/mol) 521.69
Synonym Ethanesulfonic acid, 2-[[(3α,5β,7α)-3,7-dihydroxy-24-oxocholan-24-yl]amino]-, sodium salt (1:1), Ethanesulfonic acid, 2-[[(3α,5β,7α)-3,7-dihydroxy-24-oxocholan-24-yl]amino]-, monosodium salt (9CI), Taurine, N-(3α,7α-dihydroxy-5β-cholan-24-oyl)-, monosodium salt (8CI), Taurochenodeoxycholic acid sodium salt (6CI), NSC 681055, Sodium taurochenodeoxycholate, Sodium taurochenodesoxycholate
IUPAC-namn sodium;2-[[(4R)-4-[(3R,5S,7R,8R,9S,10S,13R,14S,17R)-3,7-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]ethanesulfonate
LEDER [Na+].C[C@H](CCC(=O)NCCS(=O)(=O)[O-])[C@H]1CC[C@H]2[C@@H]3[C@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
Specifikationer
Kemiskt namn eller material Taurochenodeoxycholic Acid Sodium Salt
Anteckningar om renhetsgrad HPLC
Kvantitet 500 mg
InChI formel InChI=1S/C26H45NO6S.Na/c1-16(4-7-23(30)27-12-13-34(31,32)33)19-5-6-20-24-21(9-11-26(19,20)3)25(2)10-8-18(28)14-17(25)15-22(24)29;/h16-22,24,28-29H,4-15H2,1-3H3,(H,27,30)(H,31,32,33);/q;+1/p-1/t16-,17+,18-,19-,20+,21+,22-,24+,25+,26-;/m1./s1
Formel vikt 521.2787
Procent renhet >95%