Läs mer
Taurodeoxycholic Acid Sodium Salt, TRC
Produktkod. 17974585
Change view
Kvantitet:
100 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17974585 5 g
17941155 100 mg
17911575 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17974585 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT0090055G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1180-95-6
Molekylformel C26 H44 N O6 S . Na
Molekylvikt (g/mol) 521.69
Synonym Ethanesulfonic acid, 2-[[(3α,5β,12α)-3,12-dihydroxy-24-oxocholan-24-yl]amino]-, monosodium salt, Taurine, N-(3α,12α-dihydroxy-5β-cholan-24-oyl)-, monosodium salt (8CI), Taurodeoxycholic acid sodium salt (6CI), Cholane, ethanesulfonic acid deriv., STDC, Sodium 3α,12α-dihydroxy-5β-cholanoyltaurate, Sodium deoxytaurocholate, Sodium taurodeoxycholate, Sodium taurodesoxycholate, Taurodeoxycholate sodium
IUPAC-namn sodium;2-[[(4R)-4-[(3R,5R,8R,9S,10S,12S,13R,14S,17R)-3,12-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]ethanesulfonate
LEDER [Na+].C[C@H](CCC(=O)NCCS(=O)(=O)[O-])[C@H]1CC[C@H]2[C@@H]3CC[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3C[C@H](O)[C@]12C
Specifikationer
Kemiskt namn eller material Taurodeoxycholic Acid Sodium Salt
Anteckningar om renhetsgrad HPLC
Kvantitet 5 g
InChI formel InChI=1S/C26H45NO6S.Na/c1-16(4-9-24(30)27-12-13-34(31,32)33)20-7-8-21-19-6-5-17-14-18(28)10-11-25(17,2)22(19)15-23(29)26(20,21)3;/h16-23,28-29H,4-15H2,1-3H3,(H,27,30)(H,31,32,33);/q;+1/p-1/t16-,17-,18-,19+,20-,21+,22+,23+,25+,26-;/m1./s1
Formel vikt 521.2787
Procent renhet >95%