Läs mer
Taurolithocholic Acid Sulfate Disodium Salt, TRC
Produktkod. 30133466
Change view
Kvantitet:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133466 10 mg
30132491 100 mg
30005588 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30133466 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT00911510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 64936-83-0
Molekylformel C26 H43 N O8 S2 . 2 Na
Molekylvikt (g/mol) 607.73
Synonym 2-[[(3α,5β)-24-Oxo-3-(sulfooxy)cholan-24-yl]amino]ethanesulfonic Acid Disodium Salt, N-(3α-Hydroxy-5β-cholan-24-oyl)taurine Hydrogen Sulfate (Ester) Disodium Salt, 3α-Hydroxy-5β-cholanoyltaurine 3-Sulfate Disodium Salt, Sulfolithocholyltaurine Disodium Salt, Sulfotaurolithocholic Acid Disodium Salt, Taurolithocholic Acid 3-Sulfate Disodium Salt, Taurolithocholic Acid 3α-Sulfate Disodium Salt, Taurosulfolithocholic Acid Disodium Salt;
IUPAC-namn disodium;2-[[(4R)-4-[(3R,5R,8R,9S,10S,13R,14S,17R)-10,13-dimethyl-3-sulfonatooxy-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]ethanesulfonate
LEDER [Na+].[Na+].C[C@H](CCC(=O)NCCS(=O)(=O)[O-])[C@H]1CC[C@H]2[C@@H]3CC[C@@H]4C[C@@H](CC[C@]4(C)[C@H]3CC[C@]12C)OS(=O)(=O)[O-]
Specifikationer
Kemiskt namn eller material Taurolithocholic Acid Sulfate Disodium Salt
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C26H45NO8S2.2Na/c1-17(4-9-24(28)27-14-15-36(29,30)31)21-7-8-22-20-6-5-18-16-19(35-37(32,33)34)10-12-25(18,2)23(20)11-13-26(21,22)3;;/h17-23H,4-16H2,1-3H3,(H,27,28)(H,29,30,31)(H,32,33,34);;/q;2*+1/p-2/t17-,18-,19-,20+,21-,22+,23+,25+,26-;;/m1../s1
Formel vikt 607.2225
Procent renhet >95%