Learn More
TAX1BP3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA564090
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: PAEIAGLQIG DKIMQVNGWD MTMVTHDQAR KRLTKRSEEV VRLLVTRQSL QKAVQQSMLS Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
May regulate a number of protein-protein interactions by competing for PDZ domain binding sites. Binds CTNNB1 and may thereby act as an inhibitor of the Wnt signaling pathway. Competes with LIN7A for KCNJ4 binding, and thereby promotes KCNJ4 internalization. May play a role in the Rho signaling pathway. May play a role in activation of CDC42 by the viral protein HPV16 E6.Specifications
TAX1BP3 | |
Polyclonal | |
Unconjugated | |
TAX1BP3 | |
1300011C24Rik; C20orf100; EMBL:AAH97976.1}; GCX1; glutaminase-interacting protein 3; Tax interaction protein 1; Tax1 (human T cell leukemia virus type I) binding protein 3; Tax1 (human T-cell leukemia virus type I) binding protein 3; Tax1 binding protein 3; tax1-binding protein 3; TAX1BP3; tax1bp3 {ECO:0000312; tax-interacting protein 1; TIP1; TIP-1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
30851 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O14907 | |
TAX1BP3 | |
Recombinant protein corresponding to Human TAX1BP3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only