Läs mer
Tocris Bioscience™ Taxol Janelia Fluor 526
Beskrivning
Taxol Janelia Fluor™ 526 is a fluorogenic, green fluorescent taxol derivative for direct imaging of the microtubule cytoskeleton. Taxol Janelia Fluor™ 526 is fluorogenic: it fluoresces only once bound to microtubules, enabling hassle-free no-wash experiments. Excitation maximum = 531 nm; emission maximum = 548 nm.
Specifikationer
Specifikationer
| Kemiskt namn eller material | 4-((8-(((1 S,2 R)-3-(((2 aR,4 S,4 AS,6 R,9 S,11 S,12 S,12 aR,12 bS)-12 b-Acetoxy-12-(benzoyloxy)-4,6,11-trihydroxy-4 A,8,13,13-tetramethyl-5-oxo-2 A,3,4,4 A,5,6,9,10,11,12,12 A,12 b-dodecahydro-1 H-7,11-methanocyclodeca[3,4 ]benzo[1,2-b]oxet-9-yl)oxy)-2-hydroxy-3-oxo-1-phen |
| CAS | 2410613-93-1 |
| Kvantitet | 100 μg |
| Mål | Microtubule Probes |
| Molekylformel | C73H74F6N4O17.CF3CO2H |
| Renhet | 0.85 |