missing translation for 'onlineSavingsMsg'
Läs mer

Invitrogen™ TAZ Polyclonal Antibody

Produktkod. 15716711 Handla allt Thermo Scientific Produkter
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 μL
Förpackningsstorlek:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
15716711 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 15716711 Leverantör Invitrogen™ Leverantörsnummer PA558836

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

Denna artikel kan inte returneras. Se returpolicy

Rabbit Polyclonal Antibody

Immunogen sequence: WHVGMNDVLP NSPPYFPRFG QKITVLIGKP FSALPVLERL RAENKSAVEM RKALTDFIQE EFQHLKTQAE QLHNHLQP Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.

This gene encodes a protein that is expressed at high levels in cardiac and skeletal muscle. Mutations in this gene have been associated with a number of clinical disorders including Barth syndrome, dilated cardiomyopathy, hypertrophic DCM, endocardial fibroelastosis, and left ventricular noncompaction. Multiple transcript variants encoding different isoforms have been described. A long form and a short form of each of these isoforms is produced; the short form lacks a hydrophobic leader sequence and may exist as a cytoplasmic protein rather than being membrane-bound. Other alternatively spliced transcripts have been described but the full-length nature of all these transcripts is not known.
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen TAZ
Användningsområden Immunohistochemistry (Paraffin)
Klassificering Polyclonal
Koncentration 0.2 mg/mL
Konjugera Unconjugated
Formulering PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gen TAZ
Genaccessionsnr. Q16635
Gene Alias 2310058J06Rik; 2610021I22Rik; 5031411C02Rik; 9130012G04Rik; AW107266; AW552613; Barth syndrome); BTHS; C78399; cardiomyopathy, dilated 3A (X-linked); CMD3A; EFE; EFE2; endocardial fibroelastosis 2; FLJ27390; G4.5; HGNC:11577; LVNCX; OTTHUMP00000061673; protein G4.5; Tafazzin; tafazzin (cardiomyopathy, dilated 3A (X-linked); tafazzin (cardiomyopathy, dilated 3A (X-linked); endocardial fibroelastosis 2; Barth syndrome); Taz; Taz1; transcriptional coactivator with PDZ binding motif; transcriptional co-activator with PDZ-binding motif; transcriptional coactivator with PDZ-binding motif; transcriptional co-activator with PDZ-binding motif (TA)Z; transcriptional co-activator with PDZ-binding motif (TAZ); WW domain containing transcription regulator 1; WW domain-containing transcription regulator protein 1; Wwtr1; XAP-2; XX-FW83563B9.3
Gensymboler TAZ
Värd art Rabbit
Immunogen Recombinant protein corresponding to Human TAZ. Recombinant protein control fragment (Product #RP-95183).
Reningsmetod Antigen affinity chromatography
Kvantitet 100 μL
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 6901
Målarter Human
Innehåll och lagring Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Produkttyp Antibody
Form Liquid
Isotyp IgG
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.