Läs mer
Tazarotenic Acid Methyl Ester, TRC
Produktkod. 30134511
Change view
Kvantitet:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134511 100 mg
30137438 25 mg
30006351 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30134511 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT010065100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1332579-70-0
Molekylformel C20H19NO2S
Molekylvikt (g/mol) 337.44
Synonym 6-[2-(3,4-Dihydro-4,4-dimethyl-2H-1-benzothiopyran-6-yl)ethynyl]-,3-Pyridinecarboxylic Acid Methyl Ester;
IUPAC-namn methyl 6-[2-(4,4-dimethyl-2,3-dihydrothiochromen-6-yl)ethynyl]pyridine-3-carboxylate
LEDER COC(=O)c1ccc(nc1)C#Cc2ccc3SCCC(C)(C)c3c2
Specifikationer
Kemiskt namn eller material Tazarotenic Acid Methyl Ester
Kvantitet 100 mg
InChI formel InChI=1S/C20H19NO2S/c1-20(2)10-11-24-18-9-5-14(12-17(18)20)4-7-16-8-6-15(13-21-16)19(22)23-3/h5-6,8-9,12-13H,10-11H2,1-3H3
Formel vikt 337.1136