Läs mer
Tocris Bioscience™ TB 21007
GREENER_CHOICE
Produktkod. 18035552
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18035552 10 mg
18084592 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18035552 Leverantör Tocris Bioscience™ Leverantörsnummer 2905/10
α5-selective GABAA inverse agonist
GABAA receptor inverse agonist selective for the α5-subtype (Ki values are 1.6, 16, 20 and 20 nM for α5, α2, α1 and α3 subtypes respectively). Brain penetrant; enhances cognitive performance in rats in the delayed matching-to-place morris water maze test following i.p. administration.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 207306-50-1
Molekylformel C15H17NO2S3
Specifikationer
Kemiskt namn eller material TB 21007
Kvantitet 10 mg
Formel vikt 339.5
Renhet >99%
Molekylvikt (g/mol) 339.486