Learn More
TBC1D10A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA552517
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: NWDKWMAKKH KKIRLRCQKG IPPSLRGRAW QYLSGGKVKL QQNPGKFDEL DMSPGDPKWL DVIERDLHRQ FPFHEMFVSR GGHGQQDLFR VLKAYTLYRP EEGYCQAQAP IAAVLLMHMP AEQAFWCLVQ ICEKYLPGYY SEKLEAIQLD G Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.
TBC1D10A Polyclonal antibody specifically detects TBC1D10A in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
TBC1D10A | |
Polyclonal | |
Unconjugated | |
TBC1D10A | |
AC004997.C22.2; AI447804; dJ130H16.1; dJ130H16.2; EBP50-PDX interactor of 64 kDa; EBP50-PDZ interactor of 64 kD; EPI64; EPI64 protein; mFLJ00288; Rab27A-GAP-alpha; TBC1 domain family member 10A; TBC1 domain family, member 10A; TBC1D10; Tbc1d10a | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
83874 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
Lot-Specific | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BXI6 | |
TBC1D10A | |
Recombinant protein corresponding to Human TBC1D10A | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only