Learn More
TBC1D13 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA561110
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: EDHPLNPNPD SRWNTYFKDN EVLLQIDKDV RRLCPDISFF QRATDYPCLL ILDPQNEFET LRKRVEQTTL KSQTVAR Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.
TBC1D13 Polyclonal antibody specifically detects TBC1D13 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
TBC1D13 | |
Polyclonal | |
Unconjugated | |
TBC1D13 | |
2600014A06Rik; BC025586; TBC1 domain family member 13; TBC1 domain family, member 13; TBC1D13 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
499768, 54662, 70296 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8R3D1, Q9NVG8 | |
TBC1D13 | |
Recombinant protein corresponding to Human TBC1D13 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |