missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBC1D15 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | TBC1D15 |
---|---|
Concentration | 0.16 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16887104
|
Proteintech
17252-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16877104
|
Proteintech
17252-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Acts as a regulator of transcriptional activation. Plays a role in the inducible expression of cytokine genes in T-cells, especially in the induction of the IL-2.Specifications
TBC1D15 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q8TC07, Q9CXF4 | |
366896, 64786, 66687 | |
TBC1D15 Fusion Protein Ag10625 | |
Primary | |
-20°C | |
TBC1D15 |
0.16 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
GAP for RAB7, GTPase activating protein RAB7, Rab7 GAP, TBC1 domain family member 15, TBC1 domain family, member 15, TBC1D15 | |
Tbc1d15 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |