missing translation for 'onlineSavingsMsg'
Learn More

TBC1D28 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA548773

Additional Details : Weight : 0.01000kg

Product Code. 15693065

  • 4205.00 SEK / 400µL
Estimated Shipment: 07-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

TBC1D28 Polyclonal antibody specifically detects TBC1D28 in Human samples. It is validated for Western Blot.
Specifications

Specifications

TBC1D28
Polyclonal
PBS with 0.09% sodium azide
Q2M2D7
TBC1D28
KLH conjugated synthetic peptide between 48-77 amino acids from the Central region of human TBC1D28.
400 μL
Primary
Human
Antibody
IgG
Western Blot
Unconjugated
TBC1D28
TBC1 domain family member 28; TBC1 domain family, member 28; TBC1D28
Rabbit
Antigen affinity chromatography, Protein A
RUO
254272
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers