Learn More
TBC1D2B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA562662
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: VRSSQYDKYF TSSRLCGGVP KDTLELLHQK DDQILGLTSQ LERFSLEKES LQQEVRTLKS KVGELNEQLG MLMETIQAKD E Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 85%.
TBC1D2B is a protein coding gene. May act as a GTPase-activating protein.Specifications
TBC1D2B | |
Polyclonal | |
Unconjugated | |
TBC1D2B | |
KIAA1055; TBC1 domain family member 2B; TBC1 domain family, member 2B; TBC1D2B | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23102 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.8 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UPU7 | |
TBC1D2B | |
Recombinant protein corresponding to Human TBC1D2B | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only