Läs mer
tBID, MedChemExpress
Produktkod. 16489596
Encompass_Preferred
Change view
Kvantitet:
1 mg
5 mg
10 mg
50 mg
100 mg
Förpackningsstorlek:
100mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16489596 50 mg -
16485767 5 mg -
16464336 100 mg 100mg
16450659 1 mg -
16413038 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Produktkod. 16489596 Leverantör Medchem Express Leverantörsnummer HY100464
MedChemExpress tBID is a selective inhibitor of homeodomain-interacting protein kinase 2 (HIPK2) with an IC50 of 0.33 μM.
tBID is a selective inhibitor of homeodomain-interacting protein kinase 2 (HIPK2) with an IC50 of 0.33 μM.
Specifikationer
Kemiskt namn eller material tBID
CAS 1639895-85-4
Färg Yellow
Anteckningar om renhetsgrad Research
Molekylformel C11H3Br4N3O2
Kvantitet 50 mg
Löslighetsinformation DMSO : 20 mg/mL (37.82 mM; Need ultrasonic)
LEDER O=C1N(C2=NC=CN2)C(C3=C(Br)C(Br)=C(Br)C(Br)=C31)=O
Molekylvikt (g/mol) 528.78
Formel vikt 528.78
Procent renhet 98.0%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Cancer-Kinase/protease